Boc-Glu-NH₂ 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-(Boc-amino)-4-carbamoylbutyric acid
H62305-03
H62305-06
AAH62305-03EA
224.36
CAD
AAH62305-03
AAH62305-06
AAH62305-MD
Boc-Glu-NH₂ 98%
Boc-Glu-NH₂
4-(Boc-amino)-4-carbamoylbutyric acid, 98%
Purity: 98%
CAS No.: 18800-74-3
Molecular Formula: C10H18N2O5
Formula Weight: 246.26
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00070081
Formula:
C₁₀H₁₈N₂O₅ MW: 246.263 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070081 CAS Number: 18800-74-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...