(S)-N-tert-Butoxycarbonylalanine methyl ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Ala-OMe, N-Boc-L-alanine methyl ester

H62271-06 H62271-14
AAH62271-06EA 83.9 CAD
AAH62271-06 AAH62271-14
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester

N-Boc-L-alanine methyl ester, 95%


Purity: 95% CAS No.: 28875-17-4 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Flash Point: 113°(235°F) Physical Form: Powder Density: 1.030 Appearance: White Melting Point: 32-35° Beilstein Reference: 1872545 Optical Rotation: -45°(c=1 in methanol) MDL No.: MFCD00038513

Formula: C9H17NO4
MW: 203.24 g/mol
Melting Pt: 32…35 °C
Density: 1.030
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038513
CAS Number: 28875-17-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR