N-Boc-3-(4-pyridyl)-D-alanine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boc-D-4-Pal-OH
H62215-03
H62215-06
AAH62215-03EA
211.72
CAD
AAH62215-03
AAH62215-06
AAH62215-MD
N-Boc-3-(4-pyridyl)-D-alanine 95%
N-Boc-3-(4-pyridyl)-D-alanine
N-Boc-3-(4-pyridyl)-D-alanine, 95%
Purity: 95%
CAS No.: 37535-58-3
Molecular Formula: C13H18N2O4
Formula Weight: 266.29
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White
Melting Point: 223-229°
Beilstein Reference: 4195478
Optical Rotation: +4.1°(c=0.6 in acetic acid)
MDL No.: MFCD00672529
Formula:
C₁₃H₁₈N₂O₄ MW: 266.29 g/mol Melting Pt: 223…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672529 CAS Number: 37535-58-3 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...