Boc-D-Asp-OBzl 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-D-aspartic acid 1-benzyl ester

H62210-03 H62210-06
AAH62210-03EA 118.34 CAD
AAH62210-03 AAH62210-06 AAH62210-MD
Boc-D-Asp-OBzl 98%
Boc-D-Asp-OBzl

N-Boc-D-aspartic acid 1-benzyl ester, 98%


Purity: 98% CAS No.: 92828-64-3 Molecular Formula: C16H21NO6 Formula Weight: 323.34 Physical Form: Powder Appearance: White MDL No.: MFCD00038264

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038264
CAS Number: 92828-64-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR