(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride
L-Aspartic acid 4-methyl ester hydrochloride, 97%
Purity: 97%
CAS No.: 16856-13-6
Molecular Formula: C5H9NO4.HCl
Formula Weight: 183.59
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 3628668
MDL No.: MFCD00038972
Formula:
C5H10ClNO4 MW: 183.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038972 CAS Number: 16856-13-6 EINECS: 240-880-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...