(S)-2,3-Diaminopropanoic acid hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Dap-OH.HCl, H-Dap-OH·HCl, L-3-Aminoalanine Hydrochloride, (S)-2,3-Diaminopropionic acid monohydrochloride
(S)-2,3-Diaminopropionic acid monohydrochloride, 95%
Purity: 95%
CAS No.: 1482-97-9
Molecular Formula: C3H8N2O2.HCl
Formula Weight: 140.57
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point: 231-233° dec
Beilstein Reference: 4263944
Optical Rotation: +24°(c=2 in 0.5 M HCl)
MDL No.: MFCD00065497
Formula:
C₃H₈N₂O₂·HCl MW: 140.6 g/mol Melting Pt: 231…233 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065497 CAS Number: 1482-97-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...