Boc-glycinamide 95%
Supplier: Thermo Scientific Chemicals
H62053-03
H62053-06
AAH62053-03EA
184.86
CAD
AAH62053-03
AAH62053-06
AAH62053-MD
Boc-glycinamide 95%
Boc-glycinamide
N-Boc-glycinamide, 95%
Purity: 95%
CAS No.: 35150-09-5
Molecular Formula: C7H14N2O3
Formula Weight: 174.20
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD08275106
Formula:
C₇H₁₄N₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD08275106 CAS Number: 35150-09-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...