(S)-2-Amino-3-phenylpropanamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe-NH₂, L-Phenylalaninamide

H62030-03 H62030-06
AAH62030-03EA 73.31 CAD
AAH62030-03 AAH62030-06
(S)-2-Amino-3-phenylpropanamide 98%
(S)-2-Amino-3-phenylpropanamide

L-Phenylalaninamide, 98%


Purity: 98% CAS No.: 5241-58-7 Molecular Formula: C9H12N2O Formula Weight: 164.20 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 6270941 MDL No.: MFCD00038146

Formula: C9H12N2O
Storage Temperature: Refrigerator
MDL Number: MFCD00038146
CAS Number: 5241-58-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR