Boc-Orn-OH 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nα-Boc-L-ornithine

H62014-06 H62014-14
AAH62014-03EA 43.54 CAD
AAH62014-03 AAH62014-06 AAH62014-14
Boc-Orn-OH 95%
Boc-Orn-OH

N^a-Boc-L-ornithine, 95%


Purity: 95% CAS No.: 21887-64-9 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 Physical Form: Powder Appearance: White Melting Point: 207-208° Beilstein Reference: 4313353 MDL No.: MFCD00076970

Formula: C₁₀H₂₀N₂O₄
Melting Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00076970
CAS Number: 21887-64-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR