3-Bromoanthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3-bromobenzoic acid

H61925-03 H61925-06
AAH61925-03EA 88.95 CAD
AAH61925-03 AAH61925-06
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid

2-Amino-3-bromobenzoic acid, 95+%


Purity: 97% CAS No.: 20776-51-6 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Flash Point: 161°(322°F) Physical Form: Solid Appearance: White to pale yellow Melting Point: 174-178° Beilstein Reference: 2936928 MDL No.: MFCD03618453

Formula: C₇H₆BrNO₂
Melting Pt: 174…178 °C
Flash Pt: 161 °C(322 °F)
Storage Temperature: Ambient
MDL Number: MFCD03618453
CAS Number: 20776-51-6

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR