2-Bromo-4-(trifluoromethyl)phenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Bromo-4-hydroxybenzotrifluoride
H61836-03
H61836-06
AAH61836-03EA
76.95
CAD
AAH61836-03
AAH61836-06
2-Bromo-4-(trifluoromethyl)phenol 98%
2-Bromo-4-(trifluoromethyl)phenol
2-Bromo-4-(trifluoromethyl)phenol, 98%
Purity: 98%
CAS No.: 81107-97-3
Molecular Formula: C7H4BrF3O
Formula Weight: 241.01
Physical Form: Liquid
Appearance: Brown
Beilstein Reference: 7702274
MDL No.: MFCD06657954
Formula:
C₇H₄BrF₃O MW: 241.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657954 CAS Number: 81107-97-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...