(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine
H61820-03
H61820-06
AAH61820-03EA
88.16
CAD
AAH61820-03
AAH61820-06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid
N-Boc-O-methyl-L-tyrosine, 98%
Purity: 98%
CAS No.: 53267-93-9
Molecular Formula: C15H21NO5
Formula Weight: 295.33
Physical Form: Solid
MDL No.: MFCD00065603
Formula:
C₁₅H₂₁NO₅ MW: 295.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065603 CAS Number: 53267-93-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...