4-Fluorophenethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

H61791-14 H61791-22
AAH61791-14EA 115.02 CAD
AAH61791-14 AAH61791-22
4-Fluorophenethylamine 97%
4-Fluorophenethylamine

2-(4-Fluorophenyl)ethylamine, 97%


Purity: 97% CAS No.: 1583-88-6 Molecular Formula: C8H10FN Formula Weight: 139.17 Flash Point: 79°(174°F) Physical Form: Liquid Density: 1.061 Refractive Index: 1.507 Appearance: Yellow Boiling Point: 50-52° UN No.: 2922 MDL No.: MFCD00134208

Formula: C₈H₁₀FN
Boiling Pt: 50…52 °C
Density: 1.061
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134208
CAS Number: 1583-88-6
EINECS: 216-435-6
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR