6-Chloropyridine-2,3-diamine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Diamino-6-chloropyridine

H61716-03 H61716-06
AAH61716-03EA 115.5 CAD
AAH61716-03 AAH61716-06
6-Chloropyridine-2,3-diamine 95%
6-Chloropyridine-2,3-diamine

2,3-Diamino-6-chloropyridine, 95%


Purity: 95% CAS No.: 40851-95-4 Molecular Formula: C5H6ClN3 Formula Weight: 143.57 Storage Temperature: -30°C to -10°C UN No.: 2811 MDL No.: MFCD00209966

Formula: C5H6ClN3
MW: 143.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00209966
CAS Number: 40851-95-4
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR