5-Bromo-2-fluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61678-14
H61678-22
AAH61678-14EA
109.81
CAD
AAH61678-14
AAH61678-22
5-Bromo-2-fluoroaniline 98%
5-Bromo-2-fluoroaniline
5-Bromo-2-fluoroaniline, 98%
Purity: 98%
CAS No.: 2924-09-6
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Physical Form: Solid
Melting Point: 27°
Boiling Point: 105°/12mm
Beilstein Reference: 1241528
UN No.: 2811
MDL No.: MFCD03094181
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 105 °C/12 mm Melting Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094181 CAS Number: 2924-09-6 UN: 2811 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...