(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61638-03
H61638-06
AAH61638-03EA
100.49
CAD
AAH61638-03
AAH61638-06
AAH61638-MD
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate
N-Boc-3-iodo-L-alanine methyl ester, 98%
Purity: 98%
CAS No.: 93267-04-0
Molecular Formula: C9H16INO4
Formula Weight: 329.13
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 50-52°
Beilstein Reference: 4422011
MDL No.: MFCD00216579
Formula:
C₉H₁₆INO₄ Melting Pt: 50…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216579 CAS Number: 93267-04-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...