N-Boc-L-threonine methyl ester 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Thr-OMe, (2S,3R)-Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate
H61545-06
H61545-14
AAH61545-06EA
109.65
CAD
AAH61545-06
AAH61545-14
N-Boc-L-threonine methyl ester 95%
N-Boc-L-threonine methyl ester
N-Boc-L-threonine methyl ester, 95%
Purity: 95%
CAS No.: 79479-07-5
Molecular Formula: C10H19NO5
Formula Weight: 233.26
Physical Form: Viscous liquid
Refractive Index: 1.4500
Appearance: Yellow
Boiling Point: 206°
MDL No.: MFCD00270479
Formula:
C₁₀H₁₉NO₅ Boiling Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00270479 CAS Number: 79479-07-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...