3-Bromo-5-methylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

H61519-03 H61519-06
AAH61519-03EA 78.21 CAD
AAH61519-03 AAH61519-06
3-Bromo-5-methylaniline 98%
3-Bromo-5-methylaniline

3-Bromo-5-methylaniline, 98%


Purity: 98% CAS No.: 74586-53-1 Molecular Formula: C7H8BrN Formula Weight: 186.05 Physical Form: Powder Appearance: Brown UN No.: 2811 MDL No.: MFCD11845553

Formula: C7H8BrN
Storage Temperature: Ambient
MDL Number: MFCD11845553
CAS Number: 74586-53-1
EINECS: 277-932-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR