(5-bromothien-2-yl)methanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-Bromothien-2-yl)methanol, 5-Bromo-2-thiophenemethanol

H61493-06 H61493-14
AAH61493-06EA 219.62 CAD
AAH61493-06 AAH61493-14
(5-bromothien-2-yl)methanol 95%
(5-bromothien-2-yl)methanol

5-Bromo-2-thiophenemethanol, 95%


Purity: 95% CAS No.: 79387-71-6 Molecular Formula: C5H5BrOS Formula Weight: 193.06 Physical Form: Liquid Appearance: Pale yellow MDL No.: MFCD06202626

Formula: C₅H₅BrOS
MW: 193.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202626
CAS Number: 79387-71-6

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR