5-Bromopyrimidine-2-carbonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61363-03
H61363-06
AAH61363-03EA
69.36
CAD
AAH61363-03
AAH61363-06
5-Bromopyrimidine-2-carbonitrile 95%
5-Bromopyrimidine-2-carbonitrile
5-Bromo-2-cyanopyrimidine, 95%
Purity: 95%
CAS No.: 38275-57-9
Molecular Formula: C5H2BrN3
Formula Weight: 183.99
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White to yellow
Beilstein Reference: 775324
UN No.: 3439
MDL No.: MFCD02940446
Formula:
C₅H₂BrN₃ Storage Temperature: Ambient |
MDL Number:
MFCD02940446 CAS Number: 38275-57-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...