N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61350-06
H61350-14
AAH61350-06EA
123.71
CAD
AAH61350-06
AAH61350-14
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 95%
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid
6-(Boc-amino)hexanoic acid, 95%
Purity: 95%
CAS No.: 6404-29-1
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 35-40°
Beilstein Reference: 2049561
MDL No.: MFCD00037798
Formula:
C₁₁H₂₁NO₄ Melting Pt: 35…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037798 CAS Number: 6404-29-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...