2-Amino-6-(trifluoromethoxy)benzothiazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

H61328-06 H61328-14
AAH61328-06EA 153.1 CAD
AAH61328-06 AAH61328-14
2-Amino-6-(trifluoromethoxy)benzothiazole 98%
2-Amino-6-(trifluoromethoxy)benzothiazole

2-Amino-6-(trifluoromethoxy)benzothiazole, 98%


Purity: 98% CAS No.: 1744-22-5 Molecular Formula: C8H5F3N2OS Formula Weight: 234.20 Physical Form: Solid Appearance: White to yellow Melting Point: 114-118° Beilstein Reference: 748759 UN No.: 2811 MDL No.: MFCD00210213

Formula: C₈H₅F₃N₂OS
MW: 234.20 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00210213
CAS Number: 1744-22-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR