N-Boc-D-serine methyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61296-06
H61296-14
AAH61296-06EA
125.14
CAD
AAH61296-06
AAH61296-14
N-Boc-D-serine methyl ester 97%
N-Boc-D-serine methyl ester
N-Boc-D-serine methyl ester, 97%
Purity: 97%
CAS No.: 95715-85-8
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.080
Refractive Index: 1.4530
Appearance: Colorless to yellow
Boiling Point: 215°
Optical Rotation: +18° (c=5 in methanol)
MDL No.: MFCD00270516
Formula:
C₉H₁₇NO₅ Boiling Pt: 215 °C Density: 1.080 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00270516 CAS Number: 95715-85-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...