2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine
H61287-03
H61287-06
AAH61287-03EA
94.48
CAD
AAH61287-03
AAH61287-06
2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine
2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%
Purity: 97%
CAS No.: 197305-97-8
Molecular Formula: C5H5F3N4
Formula Weight: 178.12
Melting Point: 99-101°
UN No.: 2811
MDL No.: MFCD00115112
Formula:
C₅H₅F₃N₄ MW: 178.118 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00115112 CAS Number: 197305-97-8 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...