Boc-D-homophenylalanine 98%
Supplier: Thermo Scientific Chemicals
H61183-03
H61183-06
AAH61183-03EA
76.47
CAD
AAH61183-03
AAH61183-06
Boc-D-homophenylalanine 98%
Boc-D-homophenylalanine
(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%
Purity: 98%
CAS No.: 82732-07-8
Molecular Formula: C15H21NO4
Formula Weight: 279.33
Physical Form: Powder
Appearance: White to off-white
Melting Point: 71-78°
Beilstein Reference: 3653505
MDL No.: MFCD00076905
Formula:
C₁₅H₂₁NO₄ Melting Pt: 71…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076905 CAS Number: 82732-07-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...