Boc-Lys(Fmoc)-OH 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

H61126-06 H61126-14
AAH61126-06EA 136.04 CAD
AAH61126-06 AAH61126-14
Boc-Lys(Fmoc)-OH 98%
Boc-Lys(Fmoc)-OH

N^a-Boc-N^e-Fmoc-L-lysine, 98%


Purity: 98% CAS No.: 84624-27-1 Molecular Formula: C26H32N2O6 Formula Weight: 468.55 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White Odor: Odorless Melting Point: 93-98° Beilstein Reference: 3576584 MDL No.: MFCD00062032

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00062032
CAS Number: 84624-27-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR