Nɑ-(tert-Butoxycarbonyl)-D-asparagine 95%

Supplier: Thermo Scientific Chemicals

H61121-06 H61121-14
AAH61121-06EA 64.15 CAD
AAH61121-06 AAH61121-14
Nɑ-(tert-Butoxycarbonyl)-D-asparagine 95%
Nalpha-(tert-Butoxycarbonyl)-D-asparagine

N^a-Boc-D-asparagine, 95%


Purity: 95% CAS No.: 75647-01-7 Molecular Formula: C9H16N2O5 Formula Weight: 232.23 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 4843040 Optical Rotation: +7.3 (c=2 in DMF) MDL No.: MFCD00065558

Formula: C₉H₁₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00065558
CAS Number: 75647-01-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR