4,5-Dihydroxyanthraquinone-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60902-MD
H60902-03
CAAAH60902-MDEA
285.98
CAD
CAAAH60902-MD
AAH60902-03
4,5-Dihydroxyanthraquinone-2-carboxylic acid 97%
4,5-Dihydroxyanthraquinone-2-carboxylic acid
Rhein, 98%
Purity: 97%
CAS No.: 478-43-3
Molecular Formula: C15H8O6
Formula Weight: 284.22
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Dark yellow
Melting Point:
Formula:
C₁₅H₈O₆ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009618 CAS Number: 478-43-3 EINECS: 207-521-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...