1,3,5-Tris(4-carboxyphenyl)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H60087-03 H60087-06
CAAAH60087-03EA 188.02 CAD
CAAAH60087-03 AAH60087-06
1,3,5-Tris(4-carboxyphenyl)benzene 97%
1,3,5-Tris(4-carboxyphenyl)benzene

1,3,5-Tri(4-carboxyphenyl)benzene, 97%


Purity: 97% CAS No.: 50446-44-1 Molecular Formula: C27H18O6 Formula Weight: 438.43 Physical Form: Solid Melting Point: 322-327° UN No.: 3077 MDL No.: MFCD10000888

Formula: C₂₇H₁₈O₆
Melting Pt: 322…327 °C
Storage Temperature: Ambient
MDL Number: MFCD10000888
CAS Number: 50446-44-1
UN: 3077
ADR: "9",III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR