5-Norbornene-2-ol (endo + exo mixture) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56994-03
H56994-14
H56994-06
CAAAH56994-03EA
47.24
CAD
CAAAH56994-03
CAAAH56994-14
AAH56994-06
5-Norbornene-2-ol (endo + exo mixture) 99%
5-Norbornene-2-ol
5-Norbornen-2-ol, mixture of endo and exo, 99%
Purity: 99%
CAS No.: 13080-90-5
Molecular Formula: C7H10O
Formula Weight: 110.15
Flash Point: 51°(124°F)
Physical Form: Solid
Melting Point: 104-106°
UN No.: 1325
MDL No.: MFCD00167566
Formula:
C₇H₁₀O Melting Pt: 104…106 °C Flash Pt: 51 °C (124 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00167566 CAS Number: 13080-90-5 EINECS: 235-987-9 UN: 1325 ADR: 4.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...