Triphos 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: (2-((Diphenylphosphino)methyl)-2-methylpropane-1,3-diyl)bis(diphenylphosphine), 1,1,1-Tris(diphenylphosphinomethyl)ethane

H37863-03 H37863-06
AAH37863-03EA 116.13 CAD
AAH37863-03 AAH37863-06
Triphos 97+%
Triphos

1,1,1-Tris(diphenylphosphinomethyl)ethane, 97+%


Purity: 97+% CAS No.: 22031-12-5 Molecular Formula: C41H39P3 Formula Weight: 624.67 Physical Form: Powder Sensitivity: Air Sensitive Appearance: White Melting Point: 99-102° MDL No.: MFCD00014084

Formula: C₄₁H₃₉P₃
MW: 624.684 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00014084
CAS Number: 22031-12-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR