1-Octanol (natural) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H36188-0D
H36188-0F
H36188-AP
AAH36188-0DEA
575.12
CAD
AAH36188-0D
AAH36188-0F
AAH36188-AP
1-Octanol (natural) 98%
1-Octanol
1-Octanol, natural, 98%
Purity: 98%
CAS No.: 111-87-5
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 0.825
Refractive Index: 1.4295
Appearance: Colorless
Odor: Sweet
Melting Point: -16°
Boiling Point: 193-195°
MDL No.: MFCD00002988
Formula:
H₃C(CH₂)₆CH₂OH Boiling Pt: 193…195 °C Melting Pt: –16 °C Density: 0.825 Flash Pt: 81 °C(178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...