Oltipraz
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione, 4-Methyl-5-(pyrazin-2-yl)-3H-1,2-dithiole-3-thione
H35120-MC
H35120-ME
AAH35120-MCEA
283.83
CAD
AAH35120-MC
AAH35120-ME
Oltipraz
Oltipraz
4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione
CAS No.: 64224-21-1
Molecular Formula: C8H6N2S3
Formula Weight: 226.34
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Orange-Red
MDL No.: MFCD00868499
Formula:
C₈H₆N₂S₃ Storage Temperature: Ambient |
MDL Number:
MFCD00868499 CAS Number: 64224-21-1 EINECS: 264-736-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...