Benzyltrimethylammonium hydroxide 40% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
047189-AE
047189-AP
AA47189-AEEA
80.49
CAD
AA47189-AE
AA47189-AP
Benzyltrimethylammonium hydroxide 40% (w/w) in aqueous solution
Benzyltrimethylammonium hydroxide
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln.
CAS No.: 100-85-6
Molecular Formula: C10H17NO
Formula Weight: 167.25
Physical Form: Liquid
Density: 1.059
Refractive Index: 1.4300
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Odor: Amine-like
Beilstein Reference: 3917256
UN No.: 3267
MDL No.: MFCD00008281
Formula:
C₁₀H₁₇NO Density: 1.059 Storage Temperature: Ambient |
MDL Number:
MFCD00008281 CAS Number: 100-85-6 EINECS: 202-895-5 UN: 3267 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...