Tellurium, Plasme Standard Solution, Specpure®, Te 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
047187-AE
047187-AP
AA47187-ADEA
138.98
CAD
AA47187-AD
AA47187-AE
AA47187-AP
Tellurium, Plasme Standard Solution, Specpure®, Te 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tellurium Standards
Tellurium, plasma standard solution, Specpure|r, Te 10,000^mg/ml
Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134062
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...