Tellurium, Plasma Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
047183-AD
047183-AE
047183-AP
AA47183-ADEA
82.73
CAD
AA47183-AD
AA47183-AE
AA47183-AP
Tellurium, Plasma Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tellurium Standards
Tellurium, plasma standard solution, Specpure|r, Te 1000^mg/ml
Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134062
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...