Ruthenium(III) chloride Ru ≥ 47.7%, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ruthenium trichloride

047182-04 047182-09
AA47182-04EA 180.12 CAD
AA47182-04 AA47182-09
Ruthenium(III) chloride Ru ≥ 47.7%, anhydrous
Ruthenium (III) chloride

Ruthenium(III) chloride, anhydrous, Ru 47.7% min


CAS No.: 10049-08-8 Molecular Formula: Cl3Ru Formula Weight: 207.43 Physical Form: Powder Density: 3.110 Sensitivity: Hygroscopic Appearance: Black Odor: Pungent Melting Point: 500° dec. UN No.: 3260 MDL No.: MFCD00011208

Formula: Cl₃Ru
MW: 207.43 g/mol
Melting Pt: >500 °C (dec.)
Density: 3.110
Storage Temperature: Ambient
MDL Number: MFCD00011208
CAS Number: 10049-08-8
EINECS: 233-167-5
UN: 3260
ADR: 8,III
Merck Index: 14,8302

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR