Chromium(II) chloride ≥99.9% (metals basis) -80 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chromium dichloride
047176-03
047176-06
047176-14
AA47176-03EA
71.5
CAD
AA47176-03
AA47176-06
AA47176-14
Chromium(II) chloride ≥99.9% (metals basis) -80 mesh
Chromium (II) chloride
Chromium(II) chloride, 99.9% (metals basis), -80 Mesh Powder.
Purity: 99.9%
CAS No.: 10049-05-5
Molecular Formula: Cl2Cr
Formula Weight: 122.9
Physical Form: -80 Mesh Powder
Density: 2.878
Sensitivity: Air and Moisture Sensitive
Appearance: Grey
Odor: Odorless
Melting Point: 824°
UN No.: 3260
MDL No.: MFCD00010947
Formula:
Cl₂Cr MW: 122.9 g/mol Melting Pt: 824 °C Density: 2.878 Storage Temperature: Ambient |
MDL Number:
MFCD00010947 CAS Number: 10049-05-5 EINECS: 233-163-3 UN: 3260 ADR: 8,III Merck Index: 14,2244 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...