Tungsten, gauze woven from wire 150 mesh woven from 0.02mm (0.0008in)

Supplier: Thermo Scientific Chemicals

047167-FL 047167-GT
AA47167-FLEA 167.48 CAD
AA47167-FL AA47167-GT
Tungsten, gauze woven from wire 150 mesh woven from 0.02mm (0.0008in)
Tungsten

Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR