Antimony, plasma standard solution, Specpure®, Sb 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
047147-AD
047147-AE
AA47147-ADEA
101.91
CAD
AA47147-AD
AA47147-AE
AA47147-AP
Antimony, plasma standard solution, Specpure®, Sb 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Antimony Standards
Antimony, plasma standard solution, Specpure|r, Sb 10,000^mg/ml
Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134030
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...