Rubidium fluoride ≥99.1% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

047144-09 047144-18
AA47144-09EA 105.43 CAD
AA47144-09 AA47144-18
Rubidium fluoride ≥99.1% (metals basis)
Rubidium fluoride

Rubidium fluoride, 99.9% (metals basis)


Purity: 99.1% CAS No.: 13446-74-7 Molecular Formula: FRb Formula Weight: 104.47 Physical Form: Crystalline Density: 3.557 Refractive Index: 1.398 Sensitivity: Hygroscopic Appearance: White Melting Point: 795° UN No.: 3288 MDL No.: MFCD00011191

Formula: FRb
MW: 104.47 g/mol
Melting Pt: 795 °C
Density: 3.557
Storage Temperature: Ambient
MDL Number: MFCD00011191
CAS Number: 13446-74-7
EINECS: 236-603-2
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR