Iron(II,III) oxide 97%, nanopowder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Magnetite, Triiron tetraoxide, Iron(II,III) oxide
047141-14
047141-22
047141-36
AA47141-14EA
78.83
CAD
AA47141-14
AA47141-22
AA47141-36
Iron(II,III) oxide 97%, nanopowder
Iron (II,III) oxide
50-100nm APS Powder, S. A. 20-50m^2/g.
Purity: 97%
CAS No.: 1317-61-9
Molecular Formula: Fe3O4
Formula Weight: 231.54
Physical Form: 50-100nm APS Powder, S. A. 20-50m2/g
Density: 5.2
Refractive Index: 3.0
Appearance: Black
Odor: Odorless
Melting Point: 1538°
MDL No.: MFCD00011010
Formula:
Fe₃O₄ MW: 231.54 g/mol Melting Pt: 1538 °C Density: 5.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011010 CAS Number: 1317-61-9 EINECS: 215-277-5 Merck Index: 14,4040 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...