Tellurium, AAS Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
47130-AE 47130-AP
AA47130-AEEA 52.93 CAD
AA47130-AE AA47130-AP
Tellurium, AAS Standard Solution, Specpure®, Te 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Tellurium Standards
Tellurium, AAS standard solution, Specpure|r, Te 1000^mg/ml

Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134062
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR