Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent

Supplier: Thermo Scientific Chemicals
Danger

047124-K2 047124-K7
AA47124-K2EA 140.9 CAD
AA47124-K2 AA47124-K7
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Sodium hydroxide

Sodium hydroxide, 3.0N Standardized Solution


CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Appearance: Colorless UN No.: 1824 MDL No.: MFCD00003548

Formula: NaOH
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR