Manganese(II) fluoride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
047109-14
047109-22
AA47109-14EA
486.64
CAD
AA47109-14
AA47109-22
Manganese(II) fluoride ≥99.99% (metals basis), Puratronic®
Manganese (II) fluoride
Manganese(II) fluoride, Puratronic, 99.99% (metals basis)
Purity: 99.99%
CAS No.: 7782-64-1
Molecular Formula: F2Mn
Formula Weight: 92.93
Physical Form: Crystalline Powder
Density: 3.98
Sensitivity: Hygroscopic
Appearance: Red
Melting Point: 856°
Solubility: Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3
UN No.: 3288
MDL No.: MFCD00016222
Formula:
F₂Mn Melting Pt: 856 °C Density: 3.98 Storage Temperature: Ambient |
MDL Number:
MFCD00016222 CAS Number: 7782-64-1 EINECS: 231-960-0 UN: 3288 ADR: 6.1,III Merck Index: 14,5729 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...