Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

047103-AD 047103-AE
AA47103-ADEA 80.5 CAD
AA47103-AD AA47103-AE AA47103-AP
Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Antimony Standards

Antimony, plasma standard solution, Specpure|r, Sb 1000^mg/ml


Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134030

Order Now

SPECIFICATIONS

CAS No. 90076-67-8
Environmentally Preferable
MDL No. MFCD00134030 MFCD00134030

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR