Potassium bis(trifluoromethylsulfonyl)imide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium bis(trifluoromethanesulphonyl)imide, Potassium bis(trifluoromethanesulfonyl)imide

046909-03 046909-06 046909-MD
AA46909-03EA 161.16 CAD
AA46909-03 AA46909-06 AA46909-MD
Potassium bis(trifluoromethylsulfonyl)imide
Potassium bis(trifluoromethylsulfonyl)imide

Potassium bis(trifluoromethylsulfonyl)imide


CAS No.: 90076-67-8 Molecular Formula: C2F6KNO4S2 Formula Weight: 319.24 Physical Form: Solid Sensitivity: Moisture Sensitive Melting Point: 198-203° UN No.: 1759 MDL No.: MFCD06200829

Formula: C₂F₆KNO₄S₂
MW: 319.234 g/mol
Melting Pt: 198…203 °C
Storage Temperature: Ambient
MDL Number: MFCD06200829
CAS Number: 90076-67-8
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR