Sodium bis(trifluoromethylsulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
046640-03
046640-06
046640-MD
AA46640-03EA
164.32
CAD
AA46640-03
AA46640-06
AA46640-MD
Sodium bis(trifluoromethylsulfonyl)imide
Sodium bis(trifluoromethylsulfonyl)imide
Sodium bis(trifluoromethylsulfonyl)imide
CAS No.: 91742-21-1
Molecular Formula: C2F6NNaO4S2
Formula Weight: 303.13
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 257-258°
UN No.: 3261
MDL No.: MFCD14156021
Formula:
C₂F₆NNaO₄S₂ Melting Pt: 257…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD14156021 CAS Number: 91742-21-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...