Soldering flux, liquid

Supplier: Thermo Scientific Chemicals
Danger

045581-AE 045581-AP
AA45581-AEEA 34.79 CAD
AA45581-AE AA45581-AP
Soldering flux, liquid
Soldering flux

All purpose flux for general soldering.


Physical Form: All purpose flux for general soldering Density: 1.26 UN No.: 1840 MDL No.: MFCD01868519

Density: 1.26
Storage Temperature: Ambient
MDL Number: MFCD01868519
UN: 1840
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR