Dialamatic Microdispensers, Drummond
Supplier: Drummond
3-000-210 3-000-225 3-000-275 3-000-250
53506-270EA 889.46 CAD
53506-270 53506-274 53506-278 53507-378
Dialamatic Microdispensers, Drummond
Pipettes Repetitive Pipettors
Dialamatic Microdispensers feature borosilicate glass bores with stainless steel plungers.
  • Borosilicate glass bores with stainless steel plungers (PTFE tips on all plungers 25 µl or larger)
  • Adjustable, direct reading plunger sets volume
  • Delivery scale engraved on plunger shaft
  • Fractional deliveries possible with pre-marked indexes
  • Volumetric accuracy of ±1% at full volume
The microdispensers have a volumetric accuracy of ±1% at full volume.
The direct reading plunger sets the volume, and the plunger shaft includes delivery-scale engravings.
Order Now
SPECIFICATIONS
Environmentally Preferable